Learn More
Bellco Glass, Inc. Borosil Double Surface Condenser 300MM
SDP
Supplier:  Bellco Glass, Inc. 452800726
Encompass_Preferred
Borosil Double Surface Condenser, I/C Ground Joint 24/ 29, 300 mm, CS/5Borosil Double Surface Condensers have a water jacket on the outside and through the center of the condensing tube. This is used when the liquid is very unstable like alcohol, aromatics, essential oils, etc. These condensers are widely used throughout research laboratories. Made of 3.3 borosilicate to withstand heat and temperature changes.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearLength of Height: 300mmInterchangeable Ground Joint: 24/ 29Neck Type: JointsClass/ Quality Grade: Type I, Class AQuantity Per Case: 5
Catalog No. 50-255-0036