Learn More
Bellco Glass, Inc. Borosil Erlenmeyer Conical Flasks Narrow Mouth I/C Joint 250mL
SDP
Supplier:  Bellco Glass, Inc. 452800800
Encompass_Preferred
Borosil Erlenmeyer Conical Flasks, ISO 4797, Narrow Mouth with Interchangeable Joint, 250mL, CS/10Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosil’s Narrow Mouth Conical Erlenmeyer Flasks with Interchangeable Joint comply with IS 1381(Part II), EN ISO 4797 and are manufactured to withstand the demands of the modern laboratory. Uniform wall thickness distribution makes these flasks ideal for heating. The conical form makes these flasks also suitable for mixing liquids. The easy-to-read scale and large labeling field helps to ensure for easy marking. Come equipped with interchangeable joints.Material: 3.3 Borosilicate GlassColor: ClearCapacity: 250mLInterchangeable Joint: 24/29Neck Type: StopperClass/ Quality Grade: Type IQuantity Per Case: 10
Catalog No. 50-254-9962