Learn More
Bellco Glass, Inc. Borosil Extraction Apparatus, Soxhlet, 100mL, with 250mL Flask
SDP
Supplier:  Bellco Glass, Inc. 452800754
Encompass_Preferred
Borosil Soxhlet Extraction Apparatus with Allihn Condenser, Interchangeable Joint 100mL, Borosilicate glass, CS/1 , liquid solid extraction. Material: GlassCapacity: 100 mLOverall Dimension: 20.47" L x 9.06" W x 4.72" HBorosilicate Glass 3.3Graduation Range: Type I
Catalog No. 50-255-0008