Learn More
Bellco Glass, Inc. Borosil Filtration Assembly, 1,000mL, with 300mL Funnel, 47mm Filter Size
SDP
Supplier:  Bellco Glass, Inc. 452800819
Encompass_Preferred
Borosil Vacuum Borosilicate Glass Filtration Assembly 47MM, 1L flask 300mL funnel with an aluminum clamp.SpecificationsMaterial: GlassFlask Volume: 1LFunnel Cup Volume: 300mLMembrane Holder Diameter: 47mmClamp material: AluminumOverall Dimension: 7.68" L x 7.68" W x 8.07" H
Catalog No. 50-254-9943