Learn More
Bellco Glass, Inc. Borosil Filtration Assembly, 2,000mL, with 300mL Funnel, 47mm Filter Size
SDP
Supplier:  Bellco Glass, Inc. 452800820
Encompass_Preferred
Borosil Vacuum Borosilicate Glass Filtration Assembly 47MM, 2L flask 300 ml funnel with an aluminum clamp.SpecificationsMaterial: GlassFlask Volume: 2LFunnel Cup Volume: 300mLMembrane Holder Diameter: 47mmClamp material: AluminumOverall Dimension: 11.81" L x 11.81" W x 8.27" H
Catalog No. 50-254-9942