Learn More
Bellco Glass, Inc. Borosil Filtration Flasks 3.3 Borosilicate Tubulation 1000mL
SDP
Supplier:  Bellco Glass, Inc. 452800813
Encompass_Preferred
Borosil Filtration Flasks Bolt Neck with Tubulation, 3.3 Borosilicate, 1L, CS/10Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosil’s Bolt Neck Filtering Flasks are manufactured to withstand the demands of the modern laboratory. They are blown in special molds with a design to convey maximum mechanical strength. The neck finish is developed for strength and to ensure a proper stopper fitting.Material: 3.3 Borosilicate GlassColor: ClearCapacity: 1LApprox O.D. x Height: 136mm x 227mmClass/ Quality Grade: Type IQuantity Per Case: 10
Catalog No. 50-254-9949