Learn More
Bellco Glass, Inc. Borosil Flasks, Erlenmeyer, Narrow Mouth, Ground Glass w/ Stoppers, 50mL, 19/26
SDP
Catalog No. 502549959
Encompass_Preferred
Borosil Erlenmeyer Conical Flasks, Narrow Mouth with Interchangeable Glass Stopper, 50 mL, CS/10Uniform wall thickness distribution makes these Flasks ideal for heating. The conical form makes them suitable for mixing liquid. These Flasks can be closed with a glass stopper. Material: 3.3 Borosilicate Glass Color: ClearCapacity: 50 mLClass/ Quality Grade: Type INeck Type: StopperInterchangeable Joint: 19 / 26Quantity per Case: 10
Catalog No. 50-254-9959 Supplier Bellco Glass, Inc. Supplier No. 452800803
Borosil Erlenmeyer Conical Flasks, Narrow Mouth with Interchangeable Glass Stopper, 50 mL, CS/10Uniform wall thickness distribution makes these Flasks ideal for heating. The conical form makes them suitable for mixing liquid. These Flasks can be closed with a glass stopper. Material: 3.3 Borosilicate Glass Color: ClearCapacity: 50 mLClass/ Quality Grade: Type INeck Type: StopperInterchangeable Joint: 19 / 26Quantity per Case: 10