Learn More
Bellco Glass, Inc. Borosil Flasks, Erlenmeyer, Narrow Mouth, Ground Glass w/ Stoppers, 25mL, 14/23
SDP
Supplier:  Bellco Glass, Inc. 452800802
Encompass_Preferred
Borosil Erlenmeyer Conical Flasks, Narrow Mouth with Interchangeable Glass Stopper, 25 mL, CS/10Uniform wall thickness distribution makes these Flasks ideal for heating. The conical form makes them suitable for mixing liquid. These Flasks can be closed with a glass stopper. Material: 3.3 Borosilicate Glass Color: ClearCapacity: 25 mLClass/ Quality Grade: Type INeck Type: StopperInterchangeable Joint: 14 / 23Quantity per Case: 10
Catalog No. 50-254-9960