Learn More
Bellco Glass, Inc. Borosil Flasks, Erlenmeyer, Narrow Mouth, Beaded Rim, 250mL
SDP
Catalog No. 502549971
Encompass_Preferred
Borosil Erlenmeyer Flasks, conical, narrow mouth Borosilicate 3.3 glass, uniform wall thickness, Graduated, Larger marking area, ISO 1773, CS/40 250mLSpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 250mLColor: ClearOverall Dimension: 85mm x 145mmNeck Diameter: 34mmClass/Quality Grade: Type 1
Catalog No. 50-254-9971 Supplier Bellco Glass, Inc. Supplier No. 452800791
Borosil Erlenmeyer Flasks, conical, narrow mouth Borosilicate 3.3 glass, uniform wall thickness, Graduated, Larger marking area, ISO 1773, CS/40 250mLSpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 250mLColor: ClearOverall Dimension: 85mm x 145mmNeck Diameter: 34mmClass/Quality Grade: Type 1