Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 25mL, 12/21, Bat. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801057
Encompass_Preferred
DescriptionBorosil Wide Mouth Volumetric Flask with Interchangeable Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 25mL, Borosilicate 3.3 CS/5SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 25mLStopper Size: 12/21Color: AmberTolerance: +/- 0.08Overall Dimension:11.81" L x 8.27" W x 2.56" HClass/Quality Grade: Type 1, Class A Quantity: 5/CS
Catalog No. 50-254-9705