Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Clear, Glass St., 1,000mL, Bat. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801027
Encompass_Preferred
Borosil Wide Mouth Volumetric Flask with Interchangeable Solid Glass Stopper, Accuracy as per Class A with Batch certificate, Borosilicate 3.3 Type I, 1L, CS/10Borosil Volumetric Flasks are manufactured from 3.3 borosilicate, USP Type I glass for corrosion-free performance. The flasks are individually calibrated following an automatic calibration process which comes with the highest quality standards according to EN ISO 1042 & USP (E 288). Strict quality control through all steps eliminates possible deviations from normal volume. Volumetric Glassware offers the right mix of quality and efficiency for numerous objectives.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 1LClass/ Quality Grade: Type I, Class ATolerance: 0.4 mLQuantity Per Case: 10Neck Type: Solid Glass StopperNeck Specification: GroundedStopper Size: 24 / 29Autoclavable: YesCertified and Calibrated at 20 CAccuracy as per Class A with Batch Certificate
Catalog No. 50-254-9735