Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Clear, Glass St., 250mL, 19/26, Ind. Cert
SDP
Catalog No. 502549738
Encompass_Preferred
Borosil Wide Mouth Volumetric Flask with Interchangeble Solid Glass Stopper, Accuracy as per Class A, ASTM E288 with Individual Certificate, Borosilicate 3.3 USP Type I, 250 mL, CS/10Borosil Volumetric Flasks are manufactured from 3.3 borosilicate, USP Type I glass for corrosion-free performance. The flasks are individually calibrated following an automatic calibration process which comes with the highest quality standards according to EN ISO 1042 & USP (E 288). Strict quality control through all steps eliminates possible deviations from normal volume. Volumetric Glassware offers the right mix of quality and efficiency for numerous objectives.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 250 mLClass/ Quality Grade: Type I, Class ATolerance: 0.20 mLQuantity Per Case: 10Neck Type: Solid Glass StopperNeck Specification: GroundedStopper Size: 19 / 26Autoclavable: YesCertified and Calibrated at 20 CAccuracy as per Class A, ASTM E288 with Individual Certificate
Catalog No. 50-254-9738 Supplier Bellco Glass, Inc. Supplier No. 452801024
Borosil Wide Mouth Volumetric Flask with Interchangeble Solid Glass Stopper, Accuracy as per Class A, ASTM E288 with Individual Certificate, Borosilicate 3.3 USP Type I, 250 mL, CS/10Borosil Volumetric Flasks are manufactured from 3.3 borosilicate, USP Type I glass for corrosion-free performance. The flasks are individually calibrated following an automatic calibration process which comes with the highest quality standards according to EN ISO 1042 & USP (E 288). Strict quality control through all steps eliminates possible deviations from normal volume. Volumetric Glassware offers the right mix of quality and efficiency for numerous objectives.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 250 mLClass/ Quality Grade: Type I, Class ATolerance: 0.20 mLQuantity Per Case: 10Neck Type: Solid Glass StopperNeck Specification: GroundedStopper Size: 19 / 26Autoclavable: YesCertified and Calibrated at 20 CAccuracy as per Class A, ASTM E288 with Individual Certificate