Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 10mL, 10/19, Bat. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801053
Encompass_Preferred
DescriptionBorosil Wide Mouth Volumetric Flask with Interchangeable Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 10mL, Borosilicate 3.3 CS/5SpecificationsBottle Volume: 10mLStopper Size: 10/19Color: AmberTolerance: +/- 0.08Overall Dimension: 11.81" L x 8.27" W x 2.56" HClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9709