Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Clear, Glass St., 200mL, 19/26, Bat. Cert
SDP
Catalog No. 502549741
Encompass_Preferred
Borosil Wide Mouth Volumetric Flask with Interchangeble Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 100 mL, Borosilicate 3.3SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 200mLStopper Size: 19/26Color: ClearTolerance: .2Overall Dimension: 20.87" L x 8.66" W x 3.27" HClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9741 Supplier Bellco Glass, Inc. Supplier No. 452801021
Borosil Wide Mouth Volumetric Flask with Interchangeble Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 100 mL, Borosilicate 3.3SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 200mLStopper Size: 19/26Color: ClearTolerance: .2Overall Dimension: 20.87" L x 8.66" W x 3.27" HClass/Quality Grade: Type 1, Class A