Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 200mL, 19/26, Bat. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801063
Encompass_Preferred
DescriptionBorosil Wide Mouth Volumetric Flask with Interchangeable Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 200mL, Borosilicate 3.3 CSSpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 200mLStopper Size: 19/26Color: AmberTolerance: .2Overall Dimension: 16.93" L x 12.40" W x 7.09" HClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9699