Learn More
Bellco Glass, Inc. Borosil Funnels, Short Stem, 50mm (Medium)
SDP
Supplier:  Bellco Glass, Inc. 452800831
Encompass_Preferred
Borosil Funnels, Short Stem, Plain, 60 deg 50 MM, ISO 4798, 3.3 Borosilicate glassSpecificationsMaterial: GlassFunnel Cup Volume: 50mmBowl Angle: 60 Overall Dimension: 14.57" L x 14.57" W x 8.27" HBorosilicate Glass 3.3
Catalog No. 50-254-9931