Learn More
Bellco Glass, Inc. Borosil Funnels, Short Stem, 35mm (Small)
SDP
Catalog No. 502549932
Encompass_Preferred
Borosil Funnels, Short Stem, Plain, 60 deg bowl angle, 35 MM, DIN/ISO 4798, 3.3 Borosilicate glass SpecificationsMaterial: GlassFunnel Cup Volume: 35mmBowl Angle: 60 Overall Dimension: 14.57" L x 14.57" W x 8.27" HBorosilicate Glass 3.3
Catalog No. 50-254-9932 Supplier Bellco Glass, Inc. Supplier No. 452800830
Borosil Funnels, Short Stem, Plain, 60 deg bowl angle, 35 MM, DIN/ISO 4798, 3.3 Borosilicate glass SpecificationsMaterial: GlassFunnel Cup Volume: 35mmBowl Angle: 60 Overall Dimension: 14.57" L x 14.57" W x 8.27" HBorosilicate Glass 3.3