Learn More
Bellco Glass, Inc. Borosil Glass Separating Funnel Pear Shape PTFE Stopcock I/C Glass stopper 1000mL
SDP
Supplier:  Bellco Glass, Inc. 452800844
Encompass_Preferred
Borosil Glass Separating Funnel Pear Shape, PTFE key Stopcock, Interchangeable Glass Stopper, USP Type I Borosilicate 3.3, ISO 4800, 1 L, CS/10Borosil Funnels may be used to separate solids from liquids, liquids from liquids, and occasionally for pouring something into a container. The Pear-Shaped Separating Funnels comply with IS 1575, DIN/ ISO 4800. These funnels are specifically molded and the stopcock bores, body openings, and stems are carefully aligned to facilitate smooth and even flow of liquid with fast cut-off capabilities. This version comes with a PTFE key stopcock and is pear-shaped with a stem. Material: 3.3 Borosilicate GlassColor: ClearCapacity: 1LStopper Size: 29/32Class/ Quality Grade: Type IQuantity Per Case: 10
Catalog No. 50-254-9918