Learn More
Bellco Glass, Inc. Borosil Glass Stirrer 7*255 MM Borosilicate glass
SDP
Supplier:  Bellco Glass, Inc. 452800880
Encompass_Preferred
Borosil Glass Stirring Rod, 7mm x 255mm, CS/20Borosil Glass Stirring Rods are made of polished 3.3 borosilicate glass. Material: 3.3 Borosilicate GlassColor: ClearApprox O.D. x Length: 7mm x 255mmQuantity Per Case: 20
Catalog No. 50-254-9882