Learn More
Bellco Glass, Inc. Borosil Graduated Measuring Cylinder Single Metric Pour Out ASTM Class B 1000 mL TC
SDP
Supplier:  Bellco Glass, Inc. 452800734
Encompass_Preferred
Borosil , Graduated Measuring Cylinder, Pour Out, Single Metric, Class B Accuracy as per ASTM E 1272 with Hexagonal Base, 1 L, CS/4Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class B tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 1LGraduation Range: 10- 1000mLGraduation Interval: 10mLGraduation Color: AmberTolerance: 6.0mLClass/ Quality Grade: Type I, Class BQuantity Per Case: 4
Catalog No. 50-255-0028