Learn More
Bellco Glass, Inc. Borosil Graduated Measuring Cylinder Pour Out Single Metric ASTM 5 mL Individual Certificate, TC
SDP
Catalog No. 502549819
Encompass_Preferred
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 5mL, C/S 5Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 5mLGraduation Range: 0.1- 5mLGraduation Interval: 0.1 mLGraduation Color: AmberTolerance: 0.05mLClass/ Quality Grade: Type I, Class AQuantity Per Case: 5
Catalog No. 50-254-9819 Supplier Bellco Glass, Inc. Supplier No. 452800943
Borosil, Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 5mL, C/S 5Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 5mLGraduation Range: 0.1- 5mLGraduation Interval: 0.1 mLGraduation Color: AmberTolerance: 0.05mLClass/ Quality Grade: Type I, Class AQuantity Per Case: 5