Learn More
Bellco Glass, Inc. Borosil Graduated Measuring Cylinder Pour Out Single Metric ASTM 2000mL Individual Certificate, TC
SDP
Catalog No. 502549803
Encompass_Preferred
Measuring Cylinder, 2000mL x 20.0mLBorosil , Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 2L, C/S 4Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 2LGraduation Range: 20- 2000mLGraduation Interval: 20mLGraduation Color: AmberTolerance: 6mLClass/ Quality Grade: Type I, Class AQuantity Per Case: 4
Catalog No. 50-254-9803 Supplier Bellco Glass, Inc. Supplier No. 452800959
Measuring Cylinder, 2000mL x 20.0mLBorosil , Graduated Measuring Cylinder, Pour Out, Single Metric, Class A Accuracy as per ASTM E 1272, with Individual Certificate, with Hexagonal Base, 2L, C/S 4Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class A tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability. Comes with Individual Certificate.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 2LGraduation Range: 20- 2000mLGraduation Interval: 20mLGraduation Color: AmberTolerance: 6mLClass/ Quality Grade: Type I, Class AQuantity Per Case: 4