Learn More
Bellco Glass, Inc. Borosil Graduated Measuring Cylinder Single Metric Pour Out ASTM Class B 100 mL TC
SDP
Supplier:  Bellco Glass, Inc. 452800731
Encompass_Preferred
Borosil , Graduated Measuring Cylinder, Pour Out, Single Metric, Class B Accuracy as per ASTM E 1272 with Hexagonal Base, 100 mL, CS/5Borosil Measuring Cylinders comply with ASTM E 1272. Calibrated as per Class B tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with Amber enamel and the Cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 100mLGraduation Range: 1- 100mLGraduation Interval: 1mLGraduation Color: AmberTolerance: 1.0mLClass/ Quality Grade: Type I, Class BQuantity Per Case: 5
Catalog No. 50-255-0031