Learn More
Bellco Glass, Inc. Borosil Graham Condenser Coiled Drip Tip I/C Inner & Outer Joint 400 MM
SDP
Supplier:  Bellco Glass, Inc. 452800721
Encompass_Preferred
Borosil Graham Condenser Coiled Distillate type, Drip Tip, Interchangeable Inner and Outer Joint, 400 mm, CS/5Borosil Graham Condensers are a coiled distillate option that have an interchangeable inner and outer joint with a drip tip. The Graham Condenser has a coiled inner condensation tube providing a larger surface area for highly efficient cooling. Typical uses are for a chemical distillation application, in which you cool and condense a gas back into a liquid. Made of 3.3 borosilicate to withstand heat and temperature changes.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearApprox Jacket Length: 400mmApprox Overall Height: 550mmNeck Type: JointsInterchangeable Socket Joint: 24/ 29Interchangeable Cone Joint: 24/ 29Class/ Quality Grade: Type I, Class AQuantity Per Case: 5
Catalog No. 50-255-0041