Learn More
Bellco Glass, Inc. Borosil Liebig Condenser with I/C Inner & Outer Joint 300 MM
SDP
Supplier:  Bellco Glass, Inc. 452800714
Encompass_Preferred
Borosil Liebig Condenser with Drip Tip, Interchangeable Inner and Outer Joint, 300 mm, CS/5Borosil Liebig Condensers have an interchangeable inner and outer joint with a drip tip. This simple design has a thin wall for the inner tube, facilitating efficient heat transfer with a low cooling capacity. Most common forms of use are distillate separation. The outer glass tube has two hose connections to assist in the passing of coolant to help attribute to the cooling of hot vapors and liquids for the extraction of solvents. Made of 3.3 borosilicate to withstand heat and temperature changes.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearApprox Jacket Length: 300mmApprox Overall Height: 450mmNeck Type: JointsInterchangeable Socket Joint: 24/ 29Interchangeable Cone Joint: 24/ 29Class/ Quality Grade: Type I, Class AQuantity Per Case: 5
Catalog No. 50-255-0048