Learn More
Bellco Glass, Inc. Borosil Low Form Griffin Beaker with Spout 10 mL ISO3820Steel
SDP
Supplier:  Bellco Glass, Inc. 452800683
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout ISO 3819 Borosilicate uniform wall thickness 10mL CS/20Borosil Low Form Griffin Beakers are the ideal container for use when stirring, mixing or heating liquids. Each beaker has controlled wall thickness at the side, radius, and base which aids in the balance between thermal resistance and mechanical strength, allowing for uniform heating throughout. The pouring spout design allows for clean and precise pouring. Easy-to-read labels and graduations make for easy identification. Due to the Beaker's high resistance to chemical attack, this collection is a match for all research, industry or educational purposes. Complies with IS 2619/ ISO 3819 and DIN 12331. *5mL & 10mL Beakers not graduated. *500mL & 1L Beakers not covered in the ISO and DIN standards.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 10mLGraduation Color: Ceramic WhiteApprox O.D. x Length: 26mm x 35mmClass/ Quality Grade: Type IQuantity Per Case: 20
Catalog No. 50-255-0079