Learn More
Bellco Glass, Inc. Borosil Petri/ Culture Dish 150MM
SDP
Supplier:  Bellco Glass, Inc. 452800751
Encompass_Preferred
Borosil 3.3 Borosilicate Petri Dishes with Covers, 100 mm, CS/100Borosil brand Dishes are designed to fulfill your wide-ranging needs from Crystalizing, Evaporating, Drying, and Storing to Petri dishes for culture, assay and micro-biological works. These Petri Dishes comply with IS 2626 and are ideal for use in routine work. These Dishes are clear and withstand repeat sterilization. Each is formed to ensure even wall thickness and consistent optical performance. Borosil brand Dishes are safe, durable and economical.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 100mmApprox O.D. x Height: 100mm x 17mmClass/ Quality Grade: Type IQuantity Per Case: 100
Catalog No. 50-255-0011