Learn More
Bellco Glass, Inc. Borosil Pipettes, Measuring (Mohr), Class A, 5.0mL x 0.05mL, Batch Cert
SDP
Catalog No. 502549687
Encompass_Preferred
DescriptionBorosil Graduated Mohr Pipette Class A Batch Certificate, EN ISO 835, 3.3 Borosilicate glass, Amber markings, graduated from zero to last graduated mark, To deliver, CS/10, 5mL (5*0.05)SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 5mLOverall Dimension: 18.70" L x 18.70" W x 6.89" HGrad. Interval: .05Tolerance: .03Class/Quality Grade: Type 1, Class A
Catalog No. 50-254-9687 Supplier Bellco Glass, Inc. Supplier No. 452801075
DescriptionBorosil Graduated Mohr Pipette Class A Batch Certificate, EN ISO 835, 3.3 Borosilicate glass, Amber markings, graduated from zero to last graduated mark, To deliver, CS/10, 5mL (5*0.05)SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 5mLOverall Dimension: 18.70" L x 18.70" W x 6.89" HGrad. Interval: .05Tolerance: .03Class/Quality Grade: Type 1, Class A