Learn More
Bellco Glass, Inc. Borosil Pipettes, Serological, Class A, 10mL x 0.1mL, Batch Cert
SDP
Catalog No. 502549649
Encompass_Preferred
Borosil Graduated Serological pipettes Class A, ASTM 1293-02, Batch Certificate, 3.3 Borosilicate, To Deliver, CS/10, 10mLSpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 10mLOverall Dimension: 18.70" L x 18.70" W x 6.89" HGrad. Interval: .1Tolerance: .03Class/Quality Grade: Type 1, Class A
Catalog No. 50-254-9649 Supplier Bellco Glass, Inc. Supplier No. 452801113
Borosil Graduated Serological pipettes Class A, ASTM 1293-02, Batch Certificate, 3.3 Borosilicate, To Deliver, CS/10, 10mLSpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 10mLOverall Dimension: 18.70" L x 18.70" W x 6.89" HGrad. Interval: .1Tolerance: .03Class/Quality Grade: Type 1, Class A