Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 1mL, Batch Cert
SDP
Supplier:  Bellco Glass, Inc. 452801123
Encompass_Preferred
Borosil Volumetric Transfer Pipette Bulb Pipette Class A Accuracy ASTM 969 USP Type I, 1mL CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 1mLOverall Dimension: 14.96" L x 14.96" W x 5.12" HClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9639