Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 50mL, Batch Cert
SDP
Catalog No. 502549617
Encompass_Preferred
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 50mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 50mLOverall Dimension: 23.62" L x 9.45" W x 3.74" HTolerance: .05mLClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9617 Supplier Bellco Glass, Inc. Supplier No. 452801145
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 50mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 50mLOverall Dimension: 23.62" L x 9.45" W x 3.74" HTolerance: .05mLClass/Quality Grade: Type 1, Class A