Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 4mL, Batch Cert
SDP
Catalog No. 502549633
Encompass_Preferred
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 4mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 4mLOverall Dimension: 18.70" L x 6.89" W x 2.95" HTolerance: .01mLClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9633 Supplier Bellco Glass, Inc. Supplier No. 452801129
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 4mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 4mLOverall Dimension: 18.70" L x 6.89" W x 2.95" HTolerance: .01mLClass/Quality Grade: Type 1, Class A