Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 25mL, Batch Cert
SDP
Catalog No. 502549623
Encompass_Preferred
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 25mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 25mLOverall Dimension: 21.65" L x 7.48" W x 3.35" HTolerance: .03mLClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9623 Supplier Bellco Glass, Inc. Supplier No. 452801139
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 25mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 25mLOverall Dimension: 21.65" L x 7.48" W x 3.35" HTolerance: .03mLClass/Quality Grade: Type 1, Class A