Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 2mL, Batch Cert
SDP
Catalog No. 502549637
Encompass_Preferred
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 2mLOverall Dimension: 14.96" L x 5.12" W x 2.17" HTolerance: .006mLClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9637 Supplier Bellco Glass, Inc. Supplier No. 452801125
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 2mLOverall Dimension: 14.96" L x 5.12" W x 2.17" HTolerance: .006mLClass/Quality Grade: Type 1, Class A