Learn More
Bellco Glass, Inc. Borosil Plastic PP Stopper For Vol. Flasks 24/29
SDP
Supplier:  Bellco Glass, Inc. 452801178
Encompass_Preferred
Borosil Plastic Polypropylene Stopper for Volumetric Flasks, Interchangeable 24/29, CS/50SpecificationsMaterial: PolypropyleneColor: ClearInterchangeable: 24/29Class/ Quality Grade: Class VINeck Type: StopperQuantity Per Case: 50
Catalog No. 50-254-9584