Learn More
Bellco Glass, Inc. Borosil Reagent Square Bottles GL80 PP Screw Cap PP Pouring Ring 5000mL
SDP
Supplier:  Bellco Glass, Inc. 452800699
Encompass_Preferred
Borosil Reagent Square Glass Bottle DIN Thread Graduated GL80 PP Screw Cap Pouring Ring, 5L, CS/1Borosil Square Reagent Bottles, graduated with a polypropylene screw cap and polypropylene pouring ring are great for long-term storage and different applications in the pharmaceutical industry. The pouring ring fitting allows for a drip free experience while the specific GL 80 screw cap ensures for containment and storage. Each bottle is chemically resistant, and all components are autoclavable and sterilizable. All standard with graduations and an enameled area for marking.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 5LGraduation Color: WhiteApprox WxBxH: 160mm x 160mm x 357mmThread Specification: GL-80Neck Type: ThreadedClass/ Quality Grade: Type IQuantity Per Case: 1
Catalog No. 50-255-0063