Learn More
Bellco Glass, Inc. Borosil Retort Base, SS Steel Rod, 12 X 600
SDP
Supplier:  Bellco Glass, Inc. 452801221
Encompass_Preferred
Borosil Retort Base SS Steel Rod is made with SS Grade 201 material.SPECIFICATIONSManufacturer: BorosilSKU/Product Code: LARS8888600Quantity: Case of 2Material: Grade 201 Stainless SteelColor: SilverSize: 12mm x 600mmShipping Dimensions: 600mm (23.62") x 67mm (2.64") x 67mm (2.64")Shipping Weight: 2.25 lbs. (1.02kg)Weight each: Approximately 1 lb.
Catalog No. 50-254-9541