Learn More
Bellco Glass, Inc. Borosil Rods, Stirring, 255mm x 9mm (Large, Wide)
SDP
Catalog No. 502549881
Encompass_Preferred
Borosil Glass Stirrer 9*255 MM Borosilicate glass CS/20SpecificationsMaterial: 3.3 Borosilicate Glass Stirrer size: 9mm x 255mmColor: ClearClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9881 Supplier Bellco Glass, Inc. Supplier No. 452800881
Borosil Glass Stirrer 9*255 MM Borosilicate glass CS/20SpecificationsMaterial: 3.3 Borosilicate Glass Stirrer size: 9mm x 255mmColor: ClearClass/Quality Grade: Type 1, Class A