Learn More
Bellco Glass, Inc. Borosil Round Bottom Flask 14/23, ISO 4797, 50 mL
SDP
Catalog No. 502549792
Encompass_Preferred
Borosil Round Bottom Boiling Flask, 14/23 Interchangeable Joint, 50 mL, CS/5Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosils Round Bottom Boiling Flasks with Interchangeable Joint comply with ISO 4797. Uniform wall thickness distribution and the round bottom shape of these flasks make them ideal vessels for heating. Generally used in the distillation and extraction processes. Large, durable matt-finished enameled area for marking.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 50mLApprox Height: 14/23Interchangeable Joint: 51mm x 90mmClass/ Quality Grade: Type IQuantity Per Case: 5
Catalog No. 50-254-9792 Supplier Bellco Glass, Inc. Supplier No. 452800970
Borosil Round Bottom Boiling Flask, 14/23 Interchangeable Joint, 50 mL, CS/5Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosils Round Bottom Boiling Flasks with Interchangeable Joint comply with ISO 4797. Uniform wall thickness distribution and the round bottom shape of these flasks make them ideal vessels for heating. Generally used in the distillation and extraction processes. Large, durable matt-finished enameled area for marking.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 50mLApprox Height: 14/23Interchangeable Joint: 51mm x 90mmClass/ Quality Grade: Type IQuantity Per Case: 5