Learn More
Bellco Glass, Inc. Borosil Round Bottom Flask 19/26, ISO 4797, 50mL
SDP
Supplier:  Bellco Glass, Inc. 452800973
Encompass_Preferred
Borosil Round Bottom Boiling Flask, 19/26 Interchangeable Joint, 50 mL, CS/5Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosils Round Bottom Boiling Flasks with Interchangeable Joint comply with ISO 4797. Uniform wall thickness distribution and the round bottom shape of these flasks make them ideal vessels for heating. Generally used in the distillation and extraction processes. Large, durable matt-finished enameled area for marking.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 50mLApprox Height: 19/26Interchangeable Joint: 51mm x 90mmClass/ Quality Grade: Type IQuantity Per Case: 5
Catalog No. 50-254-9789