Learn More
Bellco Glass, Inc. Borosil Round Bottom Flask ISO 1773 2000mL
SDP
Supplier:  Bellco Glass, Inc. 452800780
Encompass_Preferred
Borosil Round Bottom Boiling Flask as per ISO 1773, 2L, CS/2Borosil brand Flasks are designed to deliver the best combination of thermal shock resistance along with physical strength and resistance to chemical attack. Borosil’s Round Bottom Boiling Flasks comply with ISO 1773 and are manufactured to withstand the demands of the modern laboratory. Uniform wall thickness distribution makes these Flasks ideal for heating. All sizes come with a durable matt-finished enameled area for marking.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 2LApprox O.D. x Height: 166mm x 260mmApprox Neck O.D.: 42mmClass/ Quality Grade: Type IQuantity Per Case: 2
Catalog No. 50-254-9982