Learn More
Bellco Glass, Inc. Borosil Solid Penny Head Glass Stopper 45/40
SDP
Supplier:  Bellco Glass, Inc. 452800864
Encompass_Preferred
Borosil Solid Penny Head Glass Stopper, Interchangeable Ground Joint 45/40, CS/20SpecificationsMaterial: 3.3 BorosilicateColor: ClearNeck Type: StopperJoint Size: 45/40Class/ Quality Grade: Type I, Class AQuantity Per Case: 20
Catalog No. 50-254-9898