Learn More
Bellco Glass, Inc. Borosil Solid Penny Head Glass Stopper 7/16
SDP
Supplier:  Bellco Glass, Inc. 452800863
Encompass_Preferred
Borosil Solid Penny Head Glass Stopper, Interchangeable Ground Joint 7/16, CS/20SpecificationsMaterial: 3.3 BorosilicateColor: ClearNeck Type: StopperJoint Size: 7/16Class/ Quality Grade: Type I, Class AQuantity Per Case: 20
Catalog No. 50-254-9899