Learn More
Bellco Glass, Inc. Borosil Tall Form Beaker with Spout 100mL
SDP
Catalog No. 502549841
Encompass_Preferred
Borosil Beaker Griffin Tall Form with Spout ISO 3819 Borosilicate uniform wall thickness 100mL CS/40Borosil Tall Form Beakers are the ideal container for use when stirring, mixing, or heating liquids. Each beaker has uniform wall thickness distribution which aids in the balance between thermal resistance and mechanical strength, which is ideal for heating liquids. The pouring spout design allows for clean and precise pouring. Each Beaker comes graduated to assist in measuring content. All sizes come standard with a durable enameled space for marking. Complies with IS 2619/ ISO 3819 and DIN 12331. *500mL not covered in IS, ISO, and DIN standards.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 100 mLGraduation Color: WhiteApprox O.D. x Length: 48mm x 80mmClass/ Quality Grade: Type IQuantity Per Case: 40Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9841 Supplier Bellco Glass, Inc. Supplier No. 452800921
Borosil Beaker Griffin Tall Form with Spout ISO 3819 Borosilicate uniform wall thickness 100mL CS/40Borosil Tall Form Beakers are the ideal container for use when stirring, mixing, or heating liquids. Each beaker has uniform wall thickness distribution which aids in the balance between thermal resistance and mechanical strength, which is ideal for heating liquids. The pouring spout design allows for clean and precise pouring. Each Beaker comes graduated to assist in measuring content. All sizes come standard with a durable enameled space for marking. Complies with IS 2619/ ISO 3819 and DIN 12331. *500mL not covered in IS, ISO, and DIN standards.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 100 mLGraduation Color: WhiteApprox O.D. x Length: 48mm x 80mmClass/ Quality Grade: Type IQuantity Per Case: 40Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.