Learn More
Bellco Glass, Inc. Borosil Tall Form Beaker with Spout 800 ML
SDP
Supplier:  Bellco Glass, Inc. 452800688
Encompass_Preferred
Borosil Beaker Griffin Tall Form with Spout ISO 3819 Borosilicate uniform wall thickness 800mL CS/20SpecificationsMaterial: 3.3 BorosilicateColor: clearHeight: 175mm(6.89")Diameter: 90mm(3.54")Case Size: 20
Catalog No. 50-255-0074