Learn More
Bellco Glass, Inc. Borosil Tall Form Beakers with Spout DB GR 150 ML
SDP
Supplier:  Bellco Glass, Inc. 452800686
Encompass_Preferred
Borosil Beaker Griffin Tall Form with Spout ISO 3819 Borosilicate uniform wall thickness 150mL CS/40Borosil Tall Form Beakers are the ideal container for use when stirring, mixing, or heating liquids. Each beaker has uniform wall thickness distribution which aids in the balance between thermal resistance and mechanical strength, which is ideal for heating liquids. The pouring spout design allows for clean and precise pouring. Each Beaker comes graduated to assist in measuring content. All sizes come standard with a durable enameled space for marking. Complies with IS 2619/ ISO 3819 and DIN 12331. *500mL not covered in IS, ISO, and DIN standards.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 150 mLGraduation Color: WhiteApprox O.D. x Length: 54mm x 95mmClass/ Quality Grade: Type IQuantity Per Case: 40
Catalog No. 50-255-0076