Learn More
Bellco Glass, Inc. Borosil Test Tubes with Rim 10*75 Borosilicate ISO 4142, 3 mL
SDP
Supplier:  Bellco Glass, Inc. 452801188
Encompass_Preferred
Borosil Test Tubes with Rim, ISO/DIN 4142, thick walled, 3mL, CS/800Borosil Test Tubes comply with IS 2618, ISO/ DIN 4142 and are designed with a rim and a round bottom. The test tubes are made thick-walled with borosilicate glass to be mechanically resistant. These test tubes have a high thermal shock resistance. Test tubes are used to hold, mix, and heat chemical experiments.SpecificationsMaterial: 3.3 BorosilicateColor: ClearCapacity: 3mLApprox O.D. x Length: 10mm x 75mmClass/ Quality Grade: Type IQuantity Per Case: 800
Catalog No. 50-254-9574