Learn More
Bellco Glass, Inc. Borosil Test Tubes with Rim 32*200, Borosilicate ISO 4142, 100 mL
SDP
Supplier:  Bellco Glass, Inc. 452801197
Encompass_Preferred
Borosil Test Tubes with Rim, ISO/DIN 4142, thick walled, 100mL, CS/50Borosil Test Tubes comply with IS 2618, ISO/ DIN 4142 and are designed with a rim and a round bottom. The test tubes are made thick-walled with borosilicate glass to be mechanically resistant. These test tubes have a high thermal shock resistance. Test tubes are used to hold, mix and heat chemical experiments.SPECIFICATIONSManufacturer: BorosilQuantity: Case of 50SKU/Product Code: 9800U10 Material: 3.3 BorosilicateColor: ClearCapacity: 100mLSize/Dimensions;Approx O.D. x Length (EA): 32mm x 200mm (1.25 x 7.87") Weight: 10.15 lbs.Class/ Quality Grade: Type I
Catalog No. 50-254-9565