Learn More
Bellco Glass, Inc. Borosil Tubes, Culture, Flat Bottom, Clear, PTFE-Lined PP Screw Caps, 15mL
SDP
Supplier:  Bellco Glass, Inc. 452800896
Encompass_Preferred
Borosil Culture Tubes Media Flat Bottom PP cap with Liner 15 mL Borosilicate CS/100SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 15 mLColor: ClearCap Material: Polypropylene Overall Dimension: 9910007Class/Quality Grade: Type 1, Class A
Catalog No. 50-254-9866