Learn More
Bellco Glass, Inc. Borosil Tubes, Culture, Round Bottom, Amber, PTFE-Lined PP Screw Caps, 30mL
SDP
Catalog No. 502549870
Encompass_Preferred
Borosil Amber Culture Tubes Media Round Bottom PP cap with Liner 30 mL Borosilicate CS/100SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 9901010Color: AmberCap Material: Polypropylene Overall Dimension: 9901010Class/Quality Grade: Type I, Class A
Catalog No. 50-254-9870 Supplier Bellco Glass, Inc. Supplier No. 452800892
Borosil Amber Culture Tubes Media Round Bottom PP cap with Liner 30 mL Borosilicate CS/100SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 9901010Color: AmberCap Material: Polypropylene Overall Dimension: 9901010Class/Quality Grade: Type I, Class A